Learn More
Medchemexpress LLC HY-N0565B 100mg , Doxycycline (hyclate) CAS:24390-14-5 Purity:>98%
SDP
Catalog No. 502259826
Encompass_Preferred
HY-N0565B 100mg Doxycycline (hyclate) CAS: 24390-14-5 Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9826 Supplier Medchemexpress LLC Supplier No. HYN0565B100MG
HY-N0565B 100mg Doxycycline (hyclate) CAS: 24390-14-5 Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.