Learn More
Apexbio Technology LLC Doxycycline hyclate, 1g Cas# 24390-14-5
SDP
Supplier:  Apexbio Technology LLC A40521000
Encompass_Preferred
Doxycycline hyclate is an antibiotic. 24390-14-5. MFCD03427564 Doxycycline hyclate is an antibiotic [1].Doxycycline hyclate is a derivative of tetracycline and possesses the activities of anti-inflammatory and antimicrobial. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8584