Learn More
Enzo Life Sciences Doxycycline . hyclate (1g). CAS: 24390-14-5
SDP
Supplier:  Enzo Life Sciences ALX380273G001
Encompass_Preferred
Broad spectrum antibiotic and bacteriostatic. Shows antiprotozoal properties. Potent inhibitor of MMPs (matrix metalloproteinases) in vivo. Inhibits collagen synthesis. Increases and promotes smooth muscle cell adhesion. Inhibits proliferation and migration. Protects microvasculature by inhibiting plasminogen systems. Alternative name: Doxycycline . hydrochloride . hemiethanolate . hemihydrate. Purity: ≥98% (HPLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9006