Learn More
Medchemexpress LLC Doxycycline hyclate | 24390-14-5 | 512.94 | 50 G
SDP
Catalog No. 5000388772
Encompass_Preferred
Doxycycline hyclate is an orally active, broad-spectrum antibiotic and metalloproteinase (MMP) inhibitor. It exhibits antibacterial activity and anti-cancer cell proliferation activity. This compound can also be utilized in constructing gene expression regulation models.
  • Orally active and broad-spectrum metalloproteinase (MMP) inhibitor.
  • Shows antibacterial activity.
  • Exhibits anti-cancer cell proliferation activity.
  • Can be used to construct gene expression regulation models.
  • Longer activity compared to Tetracycline, allowing for once or twice daily administration.
  • Shorter time to reach peak concentration and significantly longer half-life compared to Tetracycline.
Catalog No. 50-003-88772 Supplier Medchemexpress LLC Supplier No. HYN0565B50G
Doxycycline hyclate is an orally active, broad-spectrum antibiotic and metalloproteinase (MMP) inhibitor. It exhibits antibacterial activity and anti-cancer cell proliferation activity. This compound can also be utilized in constructing gene expression regulation models.
  • Orally active and broad-spectrum metalloproteinase (MMP) inhibitor.
  • Shows antibacterial activity.
  • Exhibits anti-cancer cell proliferation activity.
  • Can be used to construct gene expression regulation models.
  • Longer activity compared to Tetracycline, allowing for once or twice daily administration.
  • Shorter time to reach peak concentration and significantly longer half-life compared to Tetracycline.