Learn More
Sigma Aldrich Fine Chemicals Biosciences Doxycycline Hyclate Antimicrobial | CAS 24390-14-5 | MDL MFCD00112304 | 25 GRAMS
Catalog No. NC2330607
Encompass
"Doxycycline hyclate | Purity: 98-101% | Mol Wt: 512.94 | CAS No: 24390-14-5 | MDL No: MFCD00112304 | 25g"Doxycycline hyclate is an antimicrobial medication that exhibits broad-spectrum activity against bacterial infections. It functions as a bacteriostatic antibiotic by inhibiting bacterial protein synthesis through binding to both 30S and 50S ribosomal subunits in susceptible microorganisms such as Gram-positive, Gram-negative, aerobic, anaerobic, and atypical bacteria. This hygroscopic, yellow, crystalline powder is highly soluble in water and DMSO. It is effective in treating respiratory, genitourinary, ocular, dermatological, and sexually transmitted diseases caused by susceptible strains. In cell culture research applications, doxycycline hyclate is commonly utilized at concentrations ranging from 0.1 to 10 μg/ml.
Catalog No. NC2330607 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D52075G
"Doxycycline hyclate | Purity: 98-101% | Mol Wt: 512.94 | CAS No: 24390-14-5 | MDL No: MFCD00112304 | 25g"Doxycycline hyclate is an antimicrobial medication that exhibits broad-spectrum activity against bacterial infections. It functions as a bacteriostatic antibiotic by inhibiting bacterial protein synthesis through binding to both 30S and 50S ribosomal subunits in susceptible microorganisms such as Gram-positive, Gram-negative, aerobic, anaerobic, and atypical bacteria. This hygroscopic, yellow, crystalline powder is highly soluble in water and DMSO. It is effective in treating respiratory, genitourinary, ocular, dermatological, and sexually transmitted diseases caused by susceptible strains. In cell culture research applications, doxycycline hyclate is commonly utilized at concentrations ranging from 0.1 to 10 μg/ml.