Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Doxycycline hyclate, 98%

Catalog No. AC446060050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446060050 5 g
AC446060250 25 g
AC446061000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC446060050 Supplier Thermo Scientific Chemicals Supplier No. 446060050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Doxycycline hyclate, Doryx, CAS # 24390-14-5, is the hyclate salt form of the broad-spectrum antibiotic known as doxycycline.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Doxycycline hyclate is a broad-spectrum bacteriostatic antibiotic of the tetracycline class
  • Doxycycline inhibits bacterial growth by preventing polypeptide formation via inhibition of at least one ribosomal subunit
  • Known to reversibly bind the 30S ribosomal subunit and inhibit formation of the aminoacyl-tRNA/mRNA-ribosome complex
  • Inhibits matrix metalloproteinases in many microorganisms

Applications

  • Effective against the bacteria that cause Lyme disease, chlamydia, rickettsia and mycoplasma; suitable for antibiotic resistance testing and study of cellular mechanisms
  • Intended for laboratory use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24390-14-5
Molecular Formula 0·5 C2H6O
Molecular Weight (g/mol) 512.94
InChI Key ISZOFWGVNMFENH-YDLUHMIOSA-N
Synonym doryx, doxycycline hyclate, doxycycline, hydrochloride, hemiethanolate, hemihydrate;
PubChem CID 131676157
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
SMILES CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Merck Index 15,3488
Solubility Information Solubility in water: soluble.
Formula Weight 512.94
Percent Purity 98%
Chemical Name or Material Doxycycline hyclate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
Recommended Storage Store locked up. Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.