Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline Hyclate, G-Biosciences
SCI_ED

Catalog No. S157229
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157229 25 g
S139757 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157229 Supplier G-Biosciences Supplier No. RC1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Doxycycline hyclate is a semisynthetic tetracycline antibiotic effective against bacteria such as Mycoplasma pneumoniae and Haemophilus influenzae.

It is also a broad-spectrum inhibitor of matrix metalloproteases (MMPs). Doxycycline hyclate is routinely used to select for cells containing the pcDNA3.1 resistance plasmid in cell line 293 at an effective concentration of 2 μg/mL. It is freely soluble in aqueous solution.

Chemical Identifiers

CAS 24390-14-5
Molecular Weight (g/mol) 512.94 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 25 g
Percent Purity 98%
Grade Molecular Biology
Chemical Name or Material Doxycycline Hyclate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.