Learn More
Medchemexpress LLC Doxycycline hyclate (Standard) | 24390-14-5 | 512.94 | 50 MG
SDP
Catalog No. 5000388558
Encompass_Preferred
Doxycycline hyclate (Standard) is the analytical standard of Doxycycline hyclate. It is an antibiotic, an orally active and broad-spectrum metalloproteinase (MMP) inhibitor, showing antibacterial activity and anti-cancer cell proliferation activity. It is intended for research and analytical applications.
  • Used as an analytical standard.
  • Reference standard supplied assay.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applicable in HPLC, GC, and MS.
  • Shows antibacterial activity.
  • Shows anti-cancer cell proliferation activity.
  • Orally active and broad-spectrum metalloproteinase (MMP) inhibitor.
Catalog No. 50-003-88558 Supplier Medchemexpress LLC Supplier No. HYN0565BR50MG
Doxycycline hyclate (Standard) is the analytical standard of Doxycycline hyclate. It is an antibiotic, an orally active and broad-spectrum metalloproteinase (MMP) inhibitor, showing antibacterial activity and anti-cancer cell proliferation activity. It is intended for research and analytical applications.
  • Used as an analytical standard.
  • Reference standard supplied assay.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applicable in HPLC, GC, and MS.
  • Shows antibacterial activity.
  • Shows anti-cancer cell proliferation activity.
  • Orally active and broad-spectrum metalloproteinase (MMP) inhibitor.