Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline Hyclate, Thermo Scientific Chemicals PROMO

Catalog No. J6057922PM
Change view
Click to view available options
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
J6057922PM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J6057922PM Supplier Thermo Scientific Chemicals Supplier No. J6057922PR

Doxycycline hyclate, Doryx, CAS # 24390-14-5, is the hyclate salt form of the broad-spectrum antibiotic known as doxycycline.

General description

  • Doxycycline hyclate is a broad-spectrum bacteriostatic antibiotic of the tetracycline class
  • Doxycycline inhibits bacterial growth by preventing polypeptide formation via inhibition of at least one ribosomal subunit
  • Known to reversibly bind the 30S ribosomal subunit and inhibit formation of the aminoacyl-tRNA/mRNA-ribosome complex
  • Inhibits matrix metalloproteinases in many microorganisms

Applications

  • Effective against the bacteria that cause Lyme disease, chlamydia, rickettsia and mycoplasma; suitable for antibiotic resistance testing and study of cellular mechanisms
  • Intended for laboratory use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24390-14-5
Molecular Formula C26H41ClN2O12
Molecular Weight (g/mol) 609.066 g/mol
MDL Number MFCD07357237
InChI Key ISZOFWGVNMFENH-YDLUHMIOSA-N
Synonym Hydramycin; Doxycycline Hydrochloride Hemiethanolate Hemihydrate
PubChem CID 131676157
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
SMILES CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl

Specifications

Type Doxycycline Hyclate
Physical Form Powder
Color Yellow
Quantity 100 g
Merck Index 14,3440
Solubility Information Soluble in Water to 50mg/ml; May require warming;Soluble in Water,methanol; Sparingly soluble in ethanol; Insoluble in chloroform and ether.
Formula Weight 512.94 g/mol (503.94 g/mol Anhydrous)
Chemical Name or Material Doxycycline hyclate
Hazard Category H302-H315-H319-H335-H361fd
Hazard Statement GHS H Statement: H302-H315-H319-H335. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
RTECSNumber QI8925000
TSCA No
Recommended Storage Keep Cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.