Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline Hyclate USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048038 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-038 500 g
18-048-041 25 g
18-048-077 25 kg
18-048-101 5 kg
18-048-102 100 g
18-048-118 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-048-038 Supplier Medisca Supplier No. 043408
Add to Cart
Add to Cart

Doxycycline Hyclate USP is a yellow crystalline powder. It is hygroscopic.

Chemical Identifiers

CAS 24390-14-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color Yellow
Quantity 500 g
Percent Purity 800 to 920 μg/mg (Doxycycline)
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.