Learn More
Medchemexpress LLC Doxycycline hydrochloride | 10592-13-9 | 480.90 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0565AR
Encompass_Preferred
Doxycycline (hydrochloride) (Standard) is the analytical standard of Doxycycline (hydrochloride) and is intended for research and analytical applications. Doxycycline hydrochloride, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. It shows antibacterial activity and anti-cancer cell proliferation activity.
  • An antibiotic.
  • Orally active.
  • Broad-spectrum metalloproteinase (MMP) inhibitor.
  • Shows antibacterial activity.
  • Shows anti-cancer cell proliferation activity.
  • Analytical standard for research and analytical applications.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-90183