Learn More
Medchemexpress LLC Doxycycline hydrochloride | 10592-13-9 | MFCD03427564 | 480.90 | 50 MG
SDP
Catalog No. 5000388892
Encompass_Preferred
Doxycycline hydrochloride (Standard) is an analytical standard primarily intended for research and analytical applications. This product functions as an antibiotic and an orally active, broad-spectrum metalloproteinase (MMP) inhibitor. It also exhibits antibacterial activity and has shown anti-cancer cell proliferation activity.
  • Analytical standard for doxycycline hydrochloride.
  • Intended for research and analytical applications.
  • Acts as an orally active and broad-spectrum metalloproteinase (MMP) inhibitor.
  • Demonstrates antibacterial activity.
  • Exhibits anti-cancer cell proliferation activity.
  • Functions as a grade of analytical standard, supplied as an assay reference.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
Catalog No. 50-003-88892 Supplier Medchemexpress LLC Supplier No. HYN0565AR50MG
Doxycycline hydrochloride (Standard) is an analytical standard primarily intended for research and analytical applications. This product functions as an antibiotic and an orally active, broad-spectrum metalloproteinase (MMP) inhibitor. It also exhibits antibacterial activity and has shown anti-cancer cell proliferation activity.
  • Analytical standard for doxycycline hydrochloride.
  • Intended for research and analytical applications.
  • Acts as an orally active and broad-spectrum metalloproteinase (MMP) inhibitor.
  • Demonstrates antibacterial activity.
  • Exhibits anti-cancer cell proliferation activity.
  • Functions as a grade of analytical standard, supplied as an assay reference.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.