Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline Hydrochloride, MP Biomedicals™

Catalog No. MP219504410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219504410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219504410 Supplier MP Biomedicals, Inc Supplier No. 0219504410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

More active biologically than the corresponding 6b-epimer hydrochloride. Doxycycline Hydrochloride, MP Biomedicals is a broad spectrum antibiotic effective against gram-positive and gram-negative bacteria. It is a derivative of oxytetracycline used to inh

Doxycycline Hydrochloride is a synthetic oxytetracycline derivative and is a member of the tetracycline antibiotics group. It shows antiprotozoal properties and is a potent inhibitor of MMPs (matrix metallo-proteinases), such as type 1 collagenase. It is used to treat a variety of infections and for other biomedical applications.

  • Treats chronic prostatitis, sinusitis, pelvic inflammatory disease, acne, rosacea, and rickettsial infections
  • Inhibits the inflammatory response of Lyme disease
  • Used in prophylaxis of anthrax (caused by Bacillus anthracis)
  • Effective against Yersinia pestis
  • Inhibits collagen synthesis
  • Used in studies on wound healing and tissue remodeling
  • Eliminates Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs
  • Eliminates Ixodes scapularis ticks
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
Synonym Biocamycin, Doxigalumicina, Doxycycline hyclate, Doxy-II, Midoxin, Novadox, Retens, Tetradox, Vibradox, Hydramycin, 2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride
PubChem CID 54706018
IUPAC Name [(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Physical Form Powder
Concentration Working Concentration: It is typically used at a concentration of 20ug/mL and is stable in media at 37°C for approximately 5 days.
Quantity 10 g
Solubility Information Soluble in water (10mg/mL, clear, yellowish to yellowish-green solution) and DMSO (50mg/mL).
Formula Weight 480.9
Grade Not Available
Recommended Storage Store at 4°C, desiccate, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.