Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Doxycycline hydrochloride, 98%, [10592-13-9], D14332 480.896 C22H25ClN2O8

Catalog No. 501408976
Encompass_Preferred

Compound: Doxycycline hydrochloride

CAS#: 10592-13-9

MDL#: MFCD03427564

Background: A semi-synthetic tetracycline-class antibiotic that inhibits bacterial protein synthesis by binding the 30S ribosomal subunit. It is widely used as a research reagent and in tetracycline-inducible gene expression systems in molecular biology.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-8976 Supplier Frontier Specialty Chemicals Supplier No. D143321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Doxycycline hydrochloride

CAS#: 10592-13-9

MDL#: MFCD03427564

Background: A semi-synthetic tetracycline-class antibiotic that inhibits bacterial protein synthesis by binding the 30S ribosomal subunit. It is widely used as a research reagent and in tetracycline-inducible gene expression systems in molecular biology.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.