Learn More
Selleck Chemical LLC Doxylamine Succinate S4240-50mg
SDP
Catalog No. 503108544
Encompass_Preferred
Doxylamine succinate competitively inhibits histamine at H1 receptors with substantial sedative and anticholinergic effects
Catalog No. 50-310-8544 Supplier Selleck Chemical LLC Supplier No. S424050MG
Doxylamine succinate competitively inhibits histamine at H1 receptors with substantial sedative and anticholinergic effects