Learn More
TARGETMOL CHEMICALS INC Doxylamine succinate 200MG
SDP
Catalog No. 502880589
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Doxylamine Succinate is a pyridine derivate histamine H1 antagonist with pronounced sedative properties. Doxylamine succinate (Decapryn) competitively blocks the histamine H1 receptor and limits the typical allergic and anaphylactic responses, including bronchoconstriction, vasodilation, increased capillary permeability, and spasmodic contraction of the gastrointestinal smooth muscle, caused by actions of histamine on bronchial and gastrointestinal smooth muscles, and on capillaries. This drug also prevents histamine-induced pain and itching of the skin and mucous membranes. Purity 100%
Catalog No. 50-288-0589 Supplier TARGETMOL CHEMICALS INC Supplier No. T1115200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Doxylamine Succinate is a pyridine derivate histamine H1 antagonist with pronounced sedative properties. Doxylamine succinate (Decapryn) competitively blocks the histamine H1 receptor and limits the typical allergic and anaphylactic responses, including bronchoconstriction, vasodilation, increased capillary permeability, and spasmodic contraction of the gastrointestinal smooth muscle, caused by actions of histamine on bronchial and gastrointestinal smooth muscles, and on capillaries. This drug also prevents histamine-induced pain and itching of the skin and mucous membranes. Purity 100%