Learn More
Medchemexpress LLC Doxylamine succinate | 562-10-7 | ≥98% | C21H28N2O5 | 5 G
SDP
Catalog No. 5000432036
Encompass_Preferred
Doxylamine succinate, with a molecular weight of 388.46, is a first-generation antihistamine that acts as a histamine (H1) receptor antagonist. It also functions as a local analgesic and an effective hypnotic agent.
  • First-generation antihistamine
  • Histamine H1 receptor antagonist
  • Effective hypnotic agent
  • Local analgesic agent
  • Targets Histamine Receptor
  • Relevant to GPCR/G Protein, Immunology/Inflammation, and Neuronal Signaling pathways
Catalog No. 50-004-32036 Supplier Medchemexpress LLC Supplier No. HYA00695G
Doxylamine succinate, with a molecular weight of 388.46, is a first-generation antihistamine that acts as a histamine (H1) receptor antagonist. It also functions as a local analgesic and an effective hypnotic agent.
  • First-generation antihistamine
  • Histamine H1 receptor antagonist
  • Effective hypnotic agent
  • Local analgesic agent
  • Targets Histamine Receptor
  • Relevant to GPCR/G Protein, Immunology/Inflammation, and Neuronal Signaling pathways