Learn More
TARGETMOL CHEMICALS INC DP-1 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580075
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DP-1 hydrochloride is a degradation product of SDC-TRAP-0063 a fragment of Ganetespib a heat shock protein 90 (HSP90) inhibitor with antitumor activity. purity: 99%
Catalog No. 50-258-0075 Supplier TARGETMOL CHEMICALS INC Supplier No. T38961L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DP-1 hydrochloride is a degradation product of SDC-TRAP-0063 a fragment of Ganetespib a heat shock protein 90 (HSP90) inhibitor with antitumor activity. purity: 99%