Learn More
Apexbio Technology LLC DiscoveryProbe™ Anti-cancer Compound Library Plus 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063353
Encompass_Preferred
DiscoveryProbe Anti-cancer Compound Library comprises 1 164 bioactive compounds targeting multiple oncological pathways with documented activity against cancers including leukemia lymphoma lung and breast cancer The library integrates compounds approved by globally recognized regulatory authorities such as FDA and EMA as well as preclinical candidates extensively documented in oncology research Compounds are selected based on clearly-defined mechanisms including kinase inhibition apoptosis induction cell-cycle regulation and epigenetic modulation Representative agents within the library include Paclitaxel Imatinib Sorafenib and Vorinostat This library is suitable for cancer biology research mechanism-of-action studies drug repurposing screening and preliminary evaluations in therapeutic development contexts
Catalog No. 50-000-63353 Supplier Apexbio Technology LLC Supplier No. L1023P0.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Anti-cancer Compound Library comprises 1 164 bioactive compounds targeting multiple oncological pathways with documented activity against cancers including leukemia lymphoma lung and breast cancer The library integrates compounds approved by globally recognized regulatory authorities such as FDA and EMA as well as preclinical candidates extensively documented in oncology research Compounds are selected based on clearly-defined mechanisms including kinase inhibition apoptosis induction cell-cycle regulation and epigenetic modulation Representative agents within the library include Paclitaxel Imatinib Sorafenib and Vorinostat This library is suitable for cancer biology research mechanism-of-action studies drug repurposing screening and preliminary evaluations in therapeutic development contexts