Learn More
Apexbio Technology LLC DiscoveryProbe™ Anti-diabetic Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063743
Encompass_Preferred
The DiscoveryProbe Anti-diabetic Compound Library comprises 110 small-molecule compounds including FDA-approved agents designed for diabetes-related biomedical research This collection targets metabolic regulatory pathways involved in diabetes such as dipeptidyl peptidase-4 (DPP-4) sodium-glucose co-transporter (SGLT) poly (ADP-ribose) polymerase (PARP) and ATP-sensitive potassium channels Representative compounds include metformin glibenclamide empagliflozin rosiglitazone and sitagliptin This library serves as a resource for investigating the molecular mechanisms of diabetes conducting potential therapeutic screening studies and developing novel diabetes-relevant research models
Catalog No. 50-000-63743 Supplier Apexbio Technology LLC Supplier No. L10460.096
The DiscoveryProbe Anti-diabetic Compound Library comprises 110 small-molecule compounds including FDA-approved agents designed for diabetes-related biomedical research This collection targets metabolic regulatory pathways involved in diabetes such as dipeptidyl peptidase-4 (DPP-4) sodium-glucose co-transporter (SGLT) poly (ADP-ribose) polymerase (PARP) and ATP-sensitive potassium channels Representative compounds include metformin glibenclamide empagliflozin rosiglitazone and sitagliptin This library serves as a resource for investigating the molecular mechanisms of diabetes conducting potential therapeutic screening studies and developing novel diabetes-relevant research models