Learn More
Apexbio Technology LLC DiscoveryProbe™ Anti-infection Compound Library Plus 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063917
Encompass_Preferred
DiscoveryProbe Anti-infection Compound Library comprises 367 bioactive small-molecule compounds with demonstrated anti-infective activities against bacterial viral fungal or parasitic pathogens Compounds selected have documented mechanisms of action covering inhibition of pathogenic replication interference with cell wall synthesis disruption of membrane integrity and modulation of microbial-specific metabolic pathways Many included agents have been clinically approved by FDA or EMA aiding translational research efforts Representative compounds such as fluconazole azithromycin ribavirin and mebendazole are encompassed The library provides a relevant tool for bio-pharmaceutical researchers conducting high-throughput screening assays mechanistic studies and identification of novel therapeutic approaches in infectious disease characterization and anti-infective drug discovery
Catalog No. 50-000-63917 Supplier Apexbio Technology LLC Supplier No. L1027P0.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Anti-infection Compound Library comprises 367 bioactive small-molecule compounds with demonstrated anti-infective activities against bacterial viral fungal or parasitic pathogens Compounds selected have documented mechanisms of action covering inhibition of pathogenic replication interference with cell wall synthesis disruption of membrane integrity and modulation of microbial-specific metabolic pathways Many included agents have been clinically approved by FDA or EMA aiding translational research efforts Representative compounds such as fluconazole azithromycin ribavirin and mebendazole are encompassed The library provides a relevant tool for bio-pharmaceutical researchers conducting high-throughput screening assays mechanistic studies and identification of novel therapeutic approaches in infectious disease characterization and anti-infective drug discovery