Learn More
Apexbio Technology LLC DiscoveryProbe™ Anti-Virus Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000062820
Encompass_Preferred
The DiscoveryProbe Anti-Virus Compound Library comprises 509 structurally diverse antiviral small molecules including FDA-approved agents and preclinical research compounds Designed specifically for antiviral research in biomedical laboratories this collection covers various mechanisms of action targeting viral enzymes such as HCV protease NS3/4a protease and host-directed targets such as MEK1/2 kinases Representative compounds include protease inhibitors and kinase-pathway modulators employed clinically or in advanced experimental stages This library serves as a resource to investigate virus-host interactions antiviral efficacy studies mechanism analyses and candidate compound screenings in virology and drug discovery
Catalog No. 50-000-62820 Supplier Apexbio Technology LLC Supplier No. L10500.096
Add to Cart
Add to Cart
Request a Quote
The DiscoveryProbe Anti-Virus Compound Library comprises 509 structurally diverse antiviral small molecules including FDA-approved agents and preclinical research compounds Designed specifically for antiviral research in biomedical laboratories this collection covers various mechanisms of action targeting viral enzymes such as HCV protease NS3/4a protease and host-directed targets such as MEK1/2 kinases Representative compounds include protease inhibitors and kinase-pathway modulators employed clinically or in advanced experimental stages This library serves as a resource to investigate virus-host interactions antiviral efficacy studies mechanism analyses and candidate compound screenings in virology and drug discovery