Learn More
Apexbio Technology LLC DiscoveryProbe™ Apoptosis Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000062928
Encompass_Preferred
DiscoveryProbe Apoptosis Compound Library consists of 643 apoptosis-associated small molecules widely documented in scientific literature and approved by authoritative institutions including FDA The library targets key proteins involved in apoptosis signaling pathways such as caspases MDM2 TNF- and p53 facilitating comprehensive studies into programmed cell death mechanisms and related pathological conditions Representative compounds in this collection include selective caspase modulators TNF- pathway inhibitors and MDM2-p53 interaction agents Researchers can leverage this specialized library for apoptosis-focused drug screening mechanistic pathway exploration and therapeutic candidate identification in disease models characterized by apoptosis dysregulation
Catalog No. 50-000-62928 Supplier Apexbio Technology LLC Supplier No. L10360.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Apoptosis Compound Library consists of 643 apoptosis-associated small molecules widely documented in scientific literature and approved by authoritative institutions including FDA The library targets key proteins involved in apoptosis signaling pathways such as caspases MDM2 TNF- and p53 facilitating comprehensive studies into programmed cell death mechanisms and related pathological conditions Representative compounds in this collection include selective caspase modulators TNF- pathway inhibitors and MDM2-p53 interaction agents Researchers can leverage this specialized library for apoptosis-focused drug screening mechanistic pathway exploration and therapeutic candidate identification in disease models characterized by apoptosis dysregulation