Learn More
Apexbio Technology LLC DiscoveryProbe™ Autophagy Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063482
Encompass_Preferred
DiscoveryProbe Autophagy Compound Library is a collection of 486 small molecule compounds carefully selected based on published evidence and experimental validation for their modulatory activity on cellular autophagy pathways Autophagy a conserved cellular process regulated by molecules such as mTOR AMPK ULK1 and Beclin-1 is involved in maintaining cellular homeostasis by mediating the turnover of intracellular components Dysregulation of autophagic pathways contributes to the progression of various human disorders including cancer neurodegeneration and metabolic diseases making this library suitable for biomedical research This library offers compounds approved by regulatory agencies including FDA EMA and CFDA alongside well-characterized experimental tool inhibitors and activators such as rapamycin chloroquine bafilomycin A1 and wortmannin for evaluating autophagic flux in cellular or animal model systems
Catalog No. 50-000-63482 Supplier Apexbio Technology LLC Supplier No. L10310.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Autophagy Compound Library is a collection of 486 small molecule compounds carefully selected based on published evidence and experimental validation for their modulatory activity on cellular autophagy pathways Autophagy a conserved cellular process regulated by molecules such as mTOR AMPK ULK1 and Beclin-1 is involved in maintaining cellular homeostasis by mediating the turnover of intracellular components Dysregulation of autophagic pathways contributes to the progression of various human disorders including cancer neurodegeneration and metabolic diseases making this library suitable for biomedical research This library offers compounds approved by regulatory agencies including FDA EMA and CFDA alongside well-characterized experimental tool inhibitors and activators such as rapamycin chloroquine bafilomycin A1 and wortmannin for evaluating autophagic flux in cellular or animal model systems