Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dp-c-4 (EGFR PARP dual-targeting PROTAC) | 00-00-0 | 98.2% | 1450.93 | C72H78ClF2N15O14 | 5 MG
SDP

Catalog No. 5000223289
Encompass_Preferred

DP-C-4 is a Cereblon-based dual PROTAC that induces simultaneous degradation of EGFR and PARP in vitro, showing dose-dependent effects in cellular assays (1-50 μM; 24 h). It is intended for research into targeted protein degradation and dual-target therapeutic strategies, supplied as a high-purity solid with specific storage and solubility recommendations.

  • High purity: 98.15%.
  • Dual-target activity: degrades EGFR and PARP simultaneously.
  • Validated in cell assay: dose-dependent degradation in SW1990 cells (1-50 μM; 24 h).
  • Appearance: light yellow to green yellow solid.
  • Solubility: DMSO 12 mg/mL; may require ultrasonic agitation.
  • Storage: -80°C under nitrogen; stock solution stable up to 6 months.

Catalog No. 50-002-23289 Supplier Medchemexpress LLC Supplier No. HY1414815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DP-C-4 is a Cereblon-based dual PROTAC that induces simultaneous degradation of EGFR and PARP in vitro, showing dose-dependent effects in cellular assays (1-50 μM; 24 h). It is intended for research into targeted protein degradation and dual-target therapeutic strategies, supplied as a high-purity solid with specific storage and solubility recommendations.

  • High purity: 98.15%.
  • Dual-target activity: degrades EGFR and PARP simultaneously.
  • Validated in cell assay: dose-dependent degradation in SW1990 cells (1-50 μM; 24 h).
  • Appearance: light yellow to green yellow solid.
  • Solubility: DMSO 12 mg/mL; may require ultrasonic agitation.
  • Storage: -80°C under nitrogen; stock solution stable up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.