Learn More
Apexbio Technology LLC DiscoveryProbe™ Epigenetics Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063668
Encompass_Preferred
DiscoveryProbe Epigenetics Compound Library comprises 328 small molecules specifically targeting known epigenetic regulators and is carefully curated for biomedical research The compounds in this collection modulate essential epigenetic targets including bromodomains histone deacetylases (HDACs) Janus kinases (JAKs) histone methyltransferases (HMTs) protein Ser/Thr phosphatases and histone acetyltransferases (HATs) Representative compounds include selective HDAC inhibitors bromodomain modulators and methyltransferase-targeting molecules Compounds are approved by recognized institutions or referenced extensively in scientific literature This library facilitates epigenetic mechanism studies target identification chemical biology research and pharmacogenomic exploration
Catalog No. 50-000-63668 Supplier Apexbio Technology LLC Supplier No. L10290.096
DiscoveryProbe Epigenetics Compound Library comprises 328 small molecules specifically targeting known epigenetic regulators and is carefully curated for biomedical research The compounds in this collection modulate essential epigenetic targets including bromodomains histone deacetylases (HDACs) Janus kinases (JAKs) histone methyltransferases (HMTs) protein Ser/Thr phosphatases and histone acetyltransferases (HATs) Representative compounds include selective HDAC inhibitors bromodomain modulators and methyltransferase-targeting molecules Compounds are approved by recognized institutions or referenced extensively in scientific literature This library facilitates epigenetic mechanism studies target identification chemical biology research and pharmacogenomic exploration