Learn More
Apexbio Technology LLC DiscoveryProbe™ GPCR Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063281
Encompass_Preferred
DiscoveryProbe GPCR Compound Library comprises 931 compounds curated specifically for G protein-coupled receptor (GPCR) research a receptor family extensively targeted in therapeutic development This collection contains structurally diverse molecules with well-defined activity toward GPCRs including representative compounds such as adenosine kinase inhibitors CXCR1 and CXCR2 antagonists cannabinoid CB2 receptor agonists and GPR119 agonists Designed in compliance with internationally recognized scientific standards the library supports experimental approaches including high-throughput screening (HTS) high-content analysis (HCA) target validation and assay development thereby serving as a practical resource for pharmacological and cell-based GPCR investigations
Catalog No. 50-000-63281 Supplier Apexbio Technology LLC Supplier No. L10250.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe GPCR Compound Library comprises 931 compounds curated specifically for G protein-coupled receptor (GPCR) research a receptor family extensively targeted in therapeutic development This collection contains structurally diverse molecules with well-defined activity toward GPCRs including representative compounds such as adenosine kinase inhibitors CXCR1 and CXCR2 antagonists cannabinoid CB2 receptor agonists and GPR119 agonists Designed in compliance with internationally recognized scientific standards the library supports experimental approaches including high-throughput screening (HTS) high-content analysis (HCA) target validation and assay development thereby serving as a practical resource for pharmacological and cell-based GPCR investigations