Learn More
Apexbio Technology LLC DiscoveryProbe™ Immunology/Inflammation Compound Library Plus 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063283
Encompass_Preferred
The DiscoveryProbe Immunology/Inflammation Compound Library comprises 295 bioactive small molecules approved by regulatory bodies such as FDA and EMA or validated through peer-reviewed publications Designed specifically for biomedical research this library targets immune and inflammatory response pathways through modulation of relevant enzymes and signaling proteins including PDE IRAK TLR COX and related pathways Representative agents in this collection include known modulators such as celecoxib (COX inhibitor) roflumilast (PDE inhibitor) and IRAK inhibitor compounds This carefully curated collection provides an essential toolkit for investigating regulatory mechanisms compound screening assays target validation and drug discovery studies in inflammation and immunological disorders
Catalog No. 50-000-63283 Supplier Apexbio Technology LLC Supplier No. L1042P0.096
The DiscoveryProbe Immunology/Inflammation Compound Library comprises 295 bioactive small molecules approved by regulatory bodies such as FDA and EMA or validated through peer-reviewed publications Designed specifically for biomedical research this library targets immune and inflammatory response pathways through modulation of relevant enzymes and signaling proteins including PDE IRAK TLR COX and related pathways Representative agents in this collection include known modulators such as celecoxib (COX inhibitor) roflumilast (PDE inhibitor) and IRAK inhibitor compounds This carefully curated collection provides an essential toolkit for investigating regulatory mechanisms compound screening assays target validation and drug discovery studies in inflammation and immunological disorders