Learn More
Apexbio Technology LLC DiscoveryProbe™ Ion Channel Compound Library Plus 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063355
Encompass_Preferred
DiscoveryProbe Ion Channel Compound Library comprises 199 compounds with defined activities targeting various ion channels These channels include NMDA receptors P2X purinergic receptors TRPV1 channels AMPA receptors ATPases and TRP channels Selected compounds have documented biological activities validated by publications and recognized approvals by FDA EMA or other authoritative institutions Representative compounds such as channel antagonists and agonists facilitate mechanistic ion channel studies and pharmacological validation The library is optimized for high-throughput screening (HTS) and high-content screening (HCS) serving as a tool for exploring ion channel modulation mechanism determination target identification and functional assays in biomedical research
Catalog No. 50-000-63355 Supplier Apexbio Technology LLC Supplier No. L1030P0.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Ion Channel Compound Library comprises 199 compounds with defined activities targeting various ion channels These channels include NMDA receptors P2X purinergic receptors TRPV1 channels AMPA receptors ATPases and TRP channels Selected compounds have documented biological activities validated by publications and recognized approvals by FDA EMA or other authoritative institutions Representative compounds such as channel antagonists and agonists facilitate mechanistic ion channel studies and pharmacological validation The library is optimized for high-throughput screening (HTS) and high-content screening (HCS) serving as a tool for exploring ion channel modulation mechanism determination target identification and functional assays in biomedical research