Learn More
Apexbio Technology LLC DiscoveryProbe™ JAK/STAT Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000062700
Encompass_Preferred
The DiscoveryProbe JAK/STAT Compound Library comprises 109 small-molecule inhibitors targeting proteins in the JAK/STAT signaling pathway a pathway closely associated with immune response inflammation and cell proliferation These chemically diverse molecules include compounds targeting JAK family kinases STAT proteins Pim kinases and EGFR examples of which are ruxolitinib tofacitinib baricitinib and fedratinib approved by regulatory agencies such as FDA Designed for biological investigations the library supports systematic high-throughput screening (HTS) and high-content screening (HCS) assays facilitating research into JAK/STAT-mediated pathological processes and drug discovery efforts in oncology immunology and inflammatory disease research
Catalog No. 50-000-62700 Supplier Apexbio Technology LLC Supplier No. L10410.096
The DiscoveryProbe JAK/STAT Compound Library comprises 109 small-molecule inhibitors targeting proteins in the JAK/STAT signaling pathway a pathway closely associated with immune response inflammation and cell proliferation These chemically diverse molecules include compounds targeting JAK family kinases STAT proteins Pim kinases and EGFR examples of which are ruxolitinib tofacitinib baricitinib and fedratinib approved by regulatory agencies such as FDA Designed for biological investigations the library supports systematic high-throughput screening (HTS) and high-content screening (HCS) assays facilitating research into JAK/STAT-mediated pathological processes and drug discovery efforts in oncology immunology and inflammatory disease research