Learn More
Apexbio Technology LLC DiscoveryProbe™ Kinase Inhibitor Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000062736
Encompass_Preferred
DiscoveryProbe Kinase Inhibitor Library is a collection of 796 small-molecule kinase inhibitors including approved agents preclinical candidates and widely studied compounds developed for biological research in kinase-related signaling pathways Representative compounds such as KN-62 AZD1152 LY2874455 and Ruxolitinib phosphate are included Kinase inhibitors block kinase enzymatic activity via competitive or allosteric binding modes modulating cell signaling cascades central to cell survival proliferation differentiation and apoptosis This library is organized for high-throughput screening (HTS) and high-content screening (HCS) assays in basic biomedical research drug discovery projects and pharmacological profiling studies
Catalog No. 50-000-62736 Supplier Apexbio Technology LLC Supplier No. L10240.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Kinase Inhibitor Library is a collection of 796 small-molecule kinase inhibitors including approved agents preclinical candidates and widely studied compounds developed for biological research in kinase-related signaling pathways Representative compounds such as KN-62 AZD1152 LY2874455 and Ruxolitinib phosphate are included Kinase inhibitors block kinase enzymatic activity via competitive or allosteric binding modes modulating cell signaling cascades central to cell survival proliferation differentiation and apoptosis This library is organized for high-throughput screening (HTS) and high-content screening (HCS) assays in basic biomedical research drug discovery projects and pharmacological profiling studies