Learn More
Apexbio Technology LLC DiscoveryProbe™ Metabolism-related Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063867
Encompass_Preferred
DiscoveryProbe Metabolism-related Compound Library contains 493 biologically active small molecules targeting diverse cellular metabolic processes These compounds are well-characterized with documented bioactivity data from agencies including FDA and EMA approvals Designed specifically for research in cellular metabolism this library encompasses inhibitors agonists or modulators aimed at metabolically relevant targets such as dehydrogenases PPAR receptors HMG-CoA reductase IDO enzymes and heat shock proteins (HSPs) Representative compounds include PPAR agonists IDO inhibitors and known HMG-CoA reductase modulators enabling researchers to dissect metabolic pathways screen for novel drug targets and explore mechanisms underlying metabolic diseases
Catalog No. 50-000-63867 Supplier Apexbio Technology LLC Supplier No. L10320.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Metabolism-related Compound Library contains 493 biologically active small molecules targeting diverse cellular metabolic processes These compounds are well-characterized with documented bioactivity data from agencies including FDA and EMA approvals Designed specifically for research in cellular metabolism this library encompasses inhibitors agonists or modulators aimed at metabolically relevant targets such as dehydrogenases PPAR receptors HMG-CoA reductase IDO enzymes and heat shock proteins (HSPs) Representative compounds include PPAR agonists IDO inhibitors and known HMG-CoA reductase modulators enabling researchers to dissect metabolic pathways screen for novel drug targets and explore mechanisms underlying metabolic diseases