Learn More
Apexbio Technology LLC DiscoveryProbe™ Neuronal Signaling Compound Library Plus 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000062900
Encompass_Preferred
DiscoveryProbe Neuronal Signaling Library comprises 556 bioactive small molecules curated for neuronal signaling research designed expressly for high-throughput and high-content screening applications in neuroscience This chemically diverse library covers modulators involved in critical neuronal pathways including agents that target -secretase serotonin 5-HT3 receptors -site amyloid precursor protein cleaving enzyme (BACE) specific Na /Ca2 exchangers and benzodiazepine receptor antagonists Representative compounds such as gamma-secretase inhibitors serotonin receptor antagonists and ion channel regulators enable systematic exploration of molecular mechanisms underlying neurological processes and diseases This library serves as an essential resource for drug discovery initiatives and fundamental research approved by research institutions pursuing mechanistic insights and therapeutic intervention strategies in neurobiology
Catalog No. 50-000-62900 Supplier Apexbio Technology LLC Supplier No. L1026P0.096
DiscoveryProbe Neuronal Signaling Library comprises 556 bioactive small molecules curated for neuronal signaling research designed expressly for high-throughput and high-content screening applications in neuroscience This chemically diverse library covers modulators involved in critical neuronal pathways including agents that target -secretase serotonin 5-HT3 receptors -site amyloid precursor protein cleaving enzyme (BACE) specific Na /Ca2 exchangers and benzodiazepine receptor antagonists Representative compounds such as gamma-secretase inhibitors serotonin receptor antagonists and ion channel regulators enable systematic exploration of molecular mechanisms underlying neurological processes and diseases This library serves as an essential resource for drug discovery initiatives and fundamental research approved by research institutions pursuing mechanistic insights and therapeutic intervention strategies in neurobiology