Learn More
Apexbio Technology LLC DiscoveryProbe™ NF-κB Signaling Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Supplier:  Apexbio Technology LLC L10440.096
Encompass_Preferred
DiscoveryProbe NF- B Signaling Library contains 73 characterized inhibitors targeting diverse molecular regulators involved in the NF- B signaling pathway NF- B transcription factors play a central mechanistic role in various cellular processes such as immune response inflammation and cell proliferation Compounds in this library including inhibitors of HDAC (e g Trichostatin A Vorinostat) I B kinase (IKK) inhibitors (e g Bay 11-7082) and modulators of regulatory proteins like PGC-1 have been extensively documented by reputable agencies and peer-reviewed research literature Designed explicitly for biomedical research this compound collection serves as a strong resource to investigate NF- B-associated disease mechanisms and facilitate drug screening particularly in cancer biology and inflammation studies
Catalog No. 50-000-62702