Learn More
Apexbio Technology LLC DiscoveryProbe™ PI3K/Akt/mTOR Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063869
Encompass_Preferred
The DiscoveryProbe PI3K/Akt/mTOR Compound Library comprises 178 small molecule inhibitors targeting PI3K/Akt/mTOR-related signaling pathways This pathway regulates processes such as cell growth proliferation survival apoptosis and metabolism in various cell types making it a frequent focus for oncology and disease mechanism research The collection encompasses compounds approved by regulatory agencies including FDA and EMA and agents studied extensively in academic and pharmaceutical research settings Representative inhibitors within this library include PI3K-selective inhibitors ATM/ATR inhibitors (DNA damage response regulators) and DNA-PK and GSK-3 targeting compounds providing comprehensive coverage for biological pathway elucidation drug discovery screening and pharmacological mechanism-of-action studies
Catalog No. 50-000-63869 Supplier Apexbio Technology LLC Supplier No. L10340.096
The DiscoveryProbe PI3K/Akt/mTOR Compound Library comprises 178 small molecule inhibitors targeting PI3K/Akt/mTOR-related signaling pathways This pathway regulates processes such as cell growth proliferation survival apoptosis and metabolism in various cell types making it a frequent focus for oncology and disease mechanism research The collection encompasses compounds approved by regulatory agencies including FDA and EMA and agents studied extensively in academic and pharmaceutical research settings Representative inhibitors within this library include PI3K-selective inhibitors ATM/ATR inhibitors (DNA damage response regulators) and DNA-PK and GSK-3 targeting compounds providing comprehensive coverage for biological pathway elucidation drug discovery screening and pharmacological mechanism-of-action studies