Learn More
Apexbio Technology LLC DiscoveryProbe™ Protease Inhibitor Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063282
Encompass_Preferred
DiscoveryProbe Protease Inhibitor Library comprises 825 characterized protease inhibitors specifically developed for use in high-throughput screening (HTS) high-content screening (HCS) and advanced chemical biology studies The compounds within this collection target various proteolytic enzyme families including Cathepsins Proteasomes Gamma Secretase Heat Shock Proteins (HSP) and Amyloid beta-cleaving enzymes Representative inhibitor classes featured in this curated library include Cathepsin inhibitors proteasome modulators and secretase inhibitors This protease-targeted compound collection facilitates systematic investigation of enzyme inhibition mechanisms aiding therapeutic identification studies and drug-target validation within biomedical research
Catalog No. 50-000-63282 Supplier Apexbio Technology LLC Supplier No. L10350.096
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe Protease Inhibitor Library comprises 825 characterized protease inhibitors specifically developed for use in high-throughput screening (HTS) high-content screening (HCS) and advanced chemical biology studies The compounds within this collection target various proteolytic enzyme families including Cathepsins Proteasomes Gamma Secretase Heat Shock Proteins (HSP) and Amyloid beta-cleaving enzymes Representative inhibitor classes featured in this curated library include Cathepsin inhibitors proteasome modulators and secretase inhibitors This protease-targeted compound collection facilitates systematic investigation of enzyme inhibition mechanisms aiding therapeutic identification studies and drug-target validation within biomedical research