Learn More
Apexbio Technology LLC DiscoveryProbe™ Stem Cell Compound Library Plus 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063742
Encompass_Preferred
DiscoveryProbe Stem Cell Compound Library comprises 169 biologically characterized compounds targeting molecular pathways integral to stem cell regulation and signaling processes This library includes representative modulators affecting critical targets such as -secretase c-RET TGF- receptor type I (ALK5) and the Wnt/ -catenin axis Designed for pharmaceutical and biomedical research the collection facilitates experimental studies including stem cell differentiation regenerative medicine exploration mechanism-of-action analyses and drug screening workflows significantly broadening research utility and translational relevance
Catalog No. 50-000-63742 Supplier Apexbio Technology LLC Supplier No. L1040P0.096
DiscoveryProbe Stem Cell Compound Library comprises 169 biologically characterized compounds targeting molecular pathways integral to stem cell regulation and signaling processes This library includes representative modulators affecting critical targets such as -secretase c-RET TGF- receptor type I (ALK5) and the Wnt/ -catenin axis Designed for pharmaceutical and biomedical research the collection facilitates experimental studies including stem cell differentiation regenerative medicine exploration mechanism-of-action analyses and drug screening workflows significantly broadening research utility and translational relevance