Learn More
Apexbio Technology LLC DiscoveryProbe™ TGF-beta/Smad Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063285
Encompass_Preferred
DiscoveryProbe TGF-beta/Smad Compound Library contains 88 compounds selected for their inhibitory activities targeting TGF- /Smad signaling pathways The library encompasses inhibitors aimed at protein kinase C (PKC) Bcr-Abl and type I TGF- receptor (ALK5) Representative compounds within this collection include recognized TGF- receptor inhibitors and Smad signaling modulators approved by regulatory bodies such as the FDA and EMA This set is designed specifically for applications in high-throughput screening (HTS) high-content screening (HCS) and mechanistic studies of TGF-beta signaling in physiological and pathological contexts including fibrosis inflammation cancer progression and immunoregulation
Catalog No. 50-000-63285 Supplier Apexbio Technology LLC Supplier No. L10450.096
DiscoveryProbe TGF-beta/Smad Compound Library contains 88 compounds selected for their inhibitory activities targeting TGF- /Smad signaling pathways The library encompasses inhibitors aimed at protein kinase C (PKC) Bcr-Abl and type I TGF- receptor (ALK5) Representative compounds within this collection include recognized TGF- receptor inhibitors and Smad signaling modulators approved by regulatory bodies such as the FDA and EMA This set is designed specifically for applications in high-throughput screening (HTS) high-content screening (HCS) and mechanistic studies of TGF-beta signaling in physiological and pathological contexts including fibrosis inflammation cancer progression and immunoregulation