Learn More
Apexbio Technology LLC DiscoveryProbe™ Ubiquitination Compound Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Catalog No. 5000063356
Encompass_Preferred
DiscoveryProbe Ubiquitination Compound Library comprises 144 small molecules curated specifically for investigating ubiquitination signaling pathways These compounds engage targets such as proteasomes mTOR inhibitor of apoptosis proteins (IAPs) deubiquitinating enzymes (DUBs) and PI3K among others Provided either as lyophilized powders or dissolved in DMSO solutions the library facilitates studies examining enzyme modulation and ubiquitin-mediated protein regulation Representative molecules in this collection include proteasome inhibitors IAP antagonists and DUB modulators Applicable in cellular assays drug screening and target validation experiments the library supports fundamental biomedical research approved for use in laboratory settings
Catalog No. 50-000-63356 Supplier Apexbio Technology LLC Supplier No. L10490.096
DiscoveryProbe Ubiquitination Compound Library comprises 144 small molecules curated specifically for investigating ubiquitination signaling pathways These compounds engage targets such as proteasomes mTOR inhibitor of apoptosis proteins (IAPs) deubiquitinating enzymes (DUBs) and PI3K among others Provided either as lyophilized powders or dissolved in DMSO solutions the library facilitates studies examining enzyme modulation and ubiquitin-mediated protein regulation Representative molecules in this collection include proteasome inhibitors IAP antagonists and DUB modulators Applicable in cellular assays drug screening and target validation experiments the library supports fundamental biomedical research approved for use in laboratory settings