Learn More
Cayman Chemical DP1 Receptr Blockng PeptID 1ea
SDP
Catalog No. 502978704
Encompass_Preferred
An internal standard for the quantification of cilastatin by GC- or LC-MS
Catalog No. 50-297-8704 Supplier Cayman Chemical Supplier No. 3016401 EA
An internal standard for the quantification of cilastatin by GC- or LC-MS