Learn More
BRANSON ULTRASONICS CORPORATION (3P) DP2300 POWDER 3X2LBS PR BX 3CS
Catalog No. NC3721420
Encompass_Preferred
Branson Ultrasonics Bransonic General Purpose Cleaner, Size: 2 lb., Format: Powder, For Use With: Used to remove general soils, fingerprints, dust, light oils, greases, Safe on metals, Glass, Plastic, Includes: General Purpose Cleaner: Powder, Recommended Applications: Safety, Volume: 2 lb.
Catalog No. NC3721420 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPN955007
Branson Ultrasonics Bransonic General Purpose Cleaner, Size: 2 lb., Format: Powder, For Use With: Used to remove general soils, fingerprints, dust, light oils, greases, Safe on metals, Glass, Plastic, Includes: General Purpose Cleaner: Powder, Recommended Applications: Safety, Volume: 2 lb.