Learn More
Diagnostic Biosystems DP3 DP3 for One-Step Deparaffinization/20 Runs
Catalog No. 502407600
Encompass_Preferred
DP3 is a blend of aqueous buffers and a paraffin solvent (orange oil). Because of these properties it can act as both a deparaffinzing agent and a reydration agent thus eliminating the requirement for transferring the paraffin tissues through a series of graded xylenes and alcohol. Deparaffinization occurs rapidly at temperatures in excess of 50 C. (Range 50 - 80 C). In contrast to other aqueous deparaffinization reagents, the paraffin is completely removed from both around and beneath the tissues.\n\nDP3 deparaffinization is compatible with Immunohistochemistry.
Catalog No. 50-240-7600 Supplier Diagnostic Biosystems Supplier No. K082
DP3 is a blend of aqueous buffers and a paraffin solvent (orange oil). Because of these properties it can act as both a deparaffinzing agent and a reydration agent thus eliminating the requirement for transferring the paraffin tissues through a series of graded xylenes and alcohol. Deparaffinization occurs rapidly at temperatures in excess of 50 C. (Range 50 - 80 C). In contrast to other aqueous deparaffinization reagents, the paraffin is completely removed from both around and beneath the tissues.\n\nDP3 deparaffinization is compatible with Immunohistochemistry.