Learn More
TARGETMOL CHEMICALS INC DPBQ 5MG
SDP
Catalog No. 502581578
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DPBQ (ZINC1620467) is a p53 activator. DPBQ could activate p53 and trigger apoptosis in a polyploid-specific manner without inhibition of topoisomerase or bind DNA. purity: 98%
Catalog No. 50-258-1578 Supplier TARGETMOL CHEMICALS INC Supplier No. T110925MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DPBQ (ZINC1620467) is a p53 activator. DPBQ could activate p53 and trigger apoptosis in a polyploid-specific manner without inhibition of topoisomerase or bind DNA. purity: 98%