Learn More
Research Products International Corp DPBS 1X Solution [Dulbecco's Phosphate Buffered Saline 1X Sterile Solution], 1 Liter
SDP
Catalog No. 501368217
Encompass_Preferred
Ready to use sterile 1x solution
Contains: 137 mM Sodium Chloride
2.7 mM Potassium Chloride
10 mM Phosphates
Does not contain Calcium or Magnesium
Catalog No. 50-136-8217 Supplier Research Products International Corp Supplier No. D221501000.0
Add to Cart
Add to Cart
Ready to use sterile 1x solution
Contains: 137 mM Sodium Chloride
2.7 mM Potassium Chloride
10 mM Phosphates
Does not contain Calcium or Magnesium