Learn More
Research Products International Corp DPBS 1X Solution, 500 Milliliters
SDP
Catalog No. 501978500
Encompass_Preferred
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.
Ready to use sterile 1X solution.
Ingredients:
Sodium Chloride: 137 mM
Potassium Chloride: 2.7 mM
Phosphate Buffer: 11.9 mM
pH: 7.4
Does not contain Calcium or Magnesium
Catalog No. 50-197-8500 Supplier Research Products International Corp Supplier No. D22150500.0
Add to Cart
Add to Cart
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.
Ready to use sterile 1X solution.
Ingredients:
Sodium Chloride: 137 mM
Potassium Chloride: 2.7 mM
Phosphate Buffer: 11.9 mM
pH: 7.4
Does not contain Calcium or Magnesium