Learn More
CAPTIVATE BIO LLC DPBS no Cal. or Mag.
Catalog No. 5000876368
Encompass_Preferred
Captivate Bio s DPBS is a balanced salt solution used for various cell culture applications like cell washing transporting cells and diluting cells for counting This formulation is configured without calcium or magnesium Manufactured in the US under ISO and GMP conditions 12 x 1000mL
Catalog No. 50-008-76368 Supplier CAPTIVATE BIO LLC Supplier No. DPBS0212
Captivate Bio s DPBS is a balanced salt solution used for various cell culture applications like cell washing transporting cells and diluting cells for counting This formulation is configured without calcium or magnesium Manufactured in the US under ISO and GMP conditions 12 x 1000mL