Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Dulbecco's Phosphate-Buffered Saline (DPBS), w: Ca and Mg (6 x 500 ml)
SDP
Catalog No. 502388601
Encompass_Preferred
DPBS, WITH: Ca and Mg. QUANTITY: 6 x 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: Dulbecco's Phosphate-Buffered Saline (DPBS) with calcium chloride and magnesium chloride. The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions. These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.
Catalog No. 50-238-8601 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0435500X6
DPBS, WITH: Ca and Mg. QUANTITY: 6 x 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: Dulbecco's Phosphate-Buffered Saline (DPBS) with calcium chloride and magnesium chloride. The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions. These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.