Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco’s Phosphate Buffered Saline, 1000 mL
Catalog No. NC1652909
Encompass
Dulbecco’s Phosphate Buffered Saline (DPBS) is intended for use in the maintenance of mammalian cells where a chemically defined balanced salt solution provides an environment that will maintain the structural and physiological integrity of cells in vitro. DPBS is commonly used in cell enumerations as a diluent, for rinsing cells and as a buffer in many chromatographic procedures. DPBS also is used to wash and resuspend cells during the dissociation process, where the presence of calcium and magnesium can inhibit trypsin activity. For this application, use DPBS Modified, which is formulated without calcium and magnesium salts.
Catalog No. NC1652909 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 59331C1000ML
Dulbecco’s Phosphate Buffered Saline (DPBS) is intended for use in the maintenance of mammalian cells where a chemically defined balanced salt solution provides an environment that will maintain the structural and physiological integrity of cells in vitro. DPBS is commonly used in cell enumerations as a diluent, for rinsing cells and as a buffer in many chromatographic procedures. DPBS also is used to wash and resuspend cells during the dissociation process, where the presence of calcium and magnesium can inhibit trypsin activity. For this application, use DPBS Modified, which is formulated without calcium and magnesium salts.