Learn More
Quality Biological Inc DPBS with TWEEN 20 without Ca & Mg, 10 Liters
SDP
Catalog No. 50146761
Encompass_Preferred
DPBS can be used to provide a buffer system to maintain cell culture media in the physiological pH range of 7.0 to 7.6. ¿ DPBS serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time. ¿ DPBS is also used as a diluent of fresh red blood cells to study the trapping of red blood cell aggregate, to wash the wells of microtiter plates, and to wash cells during preparation and serial transfer. ¿ 1X working solution is recommended ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-146-761 Supplier Quality Biological Inc Supplier No. 114154151
DPBS can be used to provide a buffer system to maintain cell culture media in the physiological pH range of 7.0 to 7.6. ¿ DPBS serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time. ¿ DPBS is also used as a diluent of fresh red blood cells to study the trapping of red blood cell aggregate, to wash the wells of microtiter plates, and to wash cells during preparation and serial transfer. ¿ 1X working solution is recommended ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months