Learn More
Medchemexpress LLC DPC-681 | 284661-68-3 | 99.7% | 669.85 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY1940010MM/1ML
Encompass_Preferred
DPC-681 is a potent and selective inhibitor of HIV protease with IC90s for wild-type HIV-1 of 4 to 40 nM.
  • Potent and selective inhibitor of HIV protease.
  • Effective against wild-type HIV-1.
  • Shows no loss in potency toward recombinant mutant HIVs with the D30N mutation.
  • Exhibits a fivefold or smaller loss in potency toward mutant variants with three to five amino acid substitutions.
  • Hepatic extraction can be saturated in dogs, leading to increased bioavailability at higher doses.
Catalog No. 50-003-84003